Search
Search
| Gene Symbol: TP63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675125.2 | XP_005675182.1 | 4 - 5 | 660 | 62 | |
| XM_005675126.3 | XP_005675183.1 | 2 - 3 | 431 | 62 | ||
| XM_005675129.3 | XP_005675186.1 | 4 - 5 | 592 | 62 | ||
| XM_005675130.3 | XP_005675187.1 | 2 - 3 | 431 | 62 | ||
| XM_005675131.3 | XP_005675188.1 | 2 - 3 | 431 | 62 | ||
| XM_018046624.1 | XP_017902113.1 | 4 - 5 | 660 | 62 | ||
| XM_018046627.1 | XP_017902116.1 | 4 - 5 | 597 | 62 | ||
| XM_018046650.1 | XP_017902139.1 | 2 - 3 | 431 | 62 | ||
| XM_018046656.1 | XP_017902145.1 | 2 - 3 | 427 | 62 | ||
Set Membership: |
|