Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699573.3 | XP_005699630.1 | 2 - 3 | 328 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |