Search
Search
| Gene Symbol: MT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013970987.2 | XP_013826441.1 | 2 - 3 | 347 | 78 | |
| GenBank mRNA | EF195236.1 | - | 2 - 3 | 96 | 78 | |
| EF471976.1 | - | 2 - 3 | 96 | 78 | ||
Set Membership: |
|