Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC22A31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061879.1 | XP_017917368.1 | 3 - 4 | 580 | 100 | |
XM_018061880.1 | XP_017917369.1 | 3 - 4 | 580 | 100 | ||
GenBank mRNA | KA326469.1 | - | 3 - 4 | 591 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |