Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HPSE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698313.2 | XP_005698370.2 | 7 - 8 | 1222 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |