Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBEGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | KA336377.1 | - | 2 - 3 | 530 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |