Search
Search

PIH1D1| Gene Symbol: PIH1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692717.3 | XP_005692774.1 | 6 - 7 | 508 | 62 | |
| XM_018062780.1 | XP_017918269.1 | 6 - 7 | 591 | 62 | ||
| GenBank mRNA | JO576977.1 | - | 4 - 5 | 542 | 62 | |
Set Membership: |
|