Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SYT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054082.1 | XP_017909571.1 | 4 - 5 | 1489 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |