Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: WDR72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685754.3 | XP_005685811.2 | 17 - 18 | 3192 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |