Search
Search

LOC102184770| Gene Symbol: LOC102184770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013977337.2 | XP_013832791.1 | 2 | 337 | 103 | |
| GenBank mRNA | KA335132.1 | - | 1 | 172 | 103 | |
Set Membership: |
|