Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172995.1 | NP_001166466.1 | 7 - 8 | 1120 | 80 | |
GenBank mRNA | AF004875.1 | - | 7 - 8 | 1120 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Cavia porcellus Probe spans exons |