Search
Search

CapaR| Gene Symbol: CapaR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275246.1 | NP_001262175.1 | 9 - 10 | 1489 | 116 | |
| NM_206418.3 | NP_996140.1 | 9 - 10 | 1484 | 116 | ||
| GenBank mRNA | AF505865.1 | - | 9 - 10 | 1666 | 116 | |
| AF522193.1 | - | 9 - 10 | 1303 | 116 | ||
Set Membership: |
|