Search
Search

Eip75B| Gene Symbol: Eip75B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168757.1 | NP_730323.1 | 1 - 2 | 813 | 60 | |
| GenBank mRNA | BT032857.1 | - | 1 - 2 | 691 | 60 | |
| BT125706.1 | - | 2 - 3 | 643 | 60 | ||
Set Membership: |
|