Search
Search

skd| Gene Symbol: skd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275205.1 | NP_001262134.1 | 5 - 6 | 1165 | 101 | |
| NM_079914.3 | NP_524653.1 | 5 - 6 | 1597 | 101 | ||
| NM_168879.3 | NP_730590.1 | 5 - 6 | 1229 | 101 | ||
| NM_168880.3 | NP_730591.2 | 5 - 6 | 1385 | 101 | ||
| GenBank mRNA | AF226855.1 | - | 5 - 6 | 1589 | 101 | |
| AF227214.1 | - | 4 - 5 | 803 | 101 | ||
| AF324425.1 | - | 5 - 6 | 1147 | 101 | ||
| BT125901.1 | - | 5 - 6 | 1147 | 101 | ||
Set Membership: |
|