Search
Search

rnh1| Gene Symbol: rnh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057967.5 | NP_477315.1 | 4 - 5 | 933 | 77 | |
| NM_206055.3 | NP_995777.1 | 3 - 4 | 876 | 77 | ||
| GenBank mRNA | AY089374.1 | - | 4 - 5 | 939 | 77 | |
Set Membership: |
|