Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01794804_g1
See other PAR-1 GE Assays ›
Gene Symbol:
par-1
Gene Name:
C-TAK1-like
Gene Aliases:
27C1, BcDNA:RH48823, CG11960, CG16701, CG30131, CG30132, CG8201, DPAR-1, DPar1, Dmel\CG8201, Dmel_CG8201, EMK, MARK, PAR-1, PAR1, Par-1, Par1, anon-WO0210402.19, dMARK, dPAR-1, dPAR1, dPar-1, dPar1, l(2)27C1, l(2)k06323, par1
Chromosome Location:
Chr.2R: 19456170 - 19485686 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0260934
Assay ID
Dm01794804_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: par-1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_206171.4
NP_995893.1
11 - 12
3135
79
NM_206177.3
NP_995899.2
11 - 12
3245
79
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
3' Most
Drosophila melanogaster
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
establishment of imaginal disc-derived wing hair orientation
regulation of protein phosphorylation
protein phosphorylation
germarium-derived oocyte fate determination
border follicle cell migration
oocyte anterior/posterior axis specification
regulation of pole plasm oskar mRNA localization
pole plasm protein localization
axis specification
oocyte microtubule cytoskeleton organization
ovarian follicle cell development
border follicle cell delamination
regulation of polarized epithelial cell differentiation
negative regulation of hippo signaling
intracellular signal transduction
pole plasm oskar mRNA localization
synaptic growth at neuromuscular junction
microtubule cytoskeleton organization involved in establishment of planar polarity
positive regulation of border follicle cell migration
protein serine/threonine kinase activity
ATP binding
tau-protein kinase activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline