Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ca-alpha1T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103419.2 | NP_001096889.1 | 18 - 19 | 4852 | 72 | |
NM_001258615.1 | NP_001245544.1 | 17 - 18 | 4777 | 72 | ||
NM_001258616.1 | NP_001245545.1 | 18 - 19 | 4837 | 72 | ||
NM_001272338.1 | NP_001259267.1 | 19 - 20 | 4891 | 72 | ||
NM_132068.3 | NP_572296.2 | 18 - 19 | 4801 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |