Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pde11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103707.2 | NP_001097177.1 | 13 - 14 | 3663 | 83 | |
NM_001201908.2 | NP_001188837.1 | 13 - 14 | 3357 | 83 | ||
NM_001201909.2 | NP_001188838.1 | 13 - 14 | 3663 | 83 | ||
NM_001299115.1 | NP_001286044.1 | 13 - 14 | 3530 | 83 | ||
NM_001299116.1 | NP_001286045.1 | 14 - 15 | 2750 | 83 | ||
NM_136041.2 | NP_609885.2 | 13 - 14 | 3416 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |