Search
Search

Tsp42Ea| Gene Symbol: Tsp42Ea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080273.3 | NP_525012.2 | 2 - 3 | 207 | 87 | |
| NM_144389.2 | NP_652646.1 | 2 - 3 | 202 | 87 | ||
| NM_165502.2 | NP_724524.1 | 2 - 3 | 196 | 87 | ||
| GenBank mRNA | AF274014.1 | - | 2 - 3 | 194 | 87 | |
| AY069049.1 | - | 2 - 3 | 194 | 87 | ||
Set Membership: |
|