Search
Search

Eip78C| Gene Symbol: Eip78C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259933.1 | NP_001246862.1 | 5 - 6 | 2349 | 79 | |
| NM_001259934.1 | NP_001246863.1 | 5 - 6 | 2205 | 79 | ||
| NM_079471.3 | NP_524195.2 | 4 - 5 | 2490 | 79 | ||
| NM_168892.3 | NP_730609.3 | 2 - 3 | 1099 | 79 | ||
Set Membership: |
|