Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm01798607_m1
          See other TRL GE Assays ›
          Gene Symbol:
          Trl CG42507
          Gene Name:
          Trithorax-like
          CG42507-PA
          Gene Aliases:
          Adf-2, Adf-2-519, Adf2, CG33261, CG9343, Dmel\CG33261, Dmel_CG33261, E(var)3-trl, E(var)62, GAF, GAGA, GAGA factor, Gaf, Gaga, LBC, NC70F, Nc70F, TRL, TfGAGA/Adf-2, Trl-GAGA, anon-EST:fe2E12, gaga, l(3)s2325, trl
          Dmel\CG42507, Dmel_CG42507
          Chromosome Location:
          Chr.3L: 14747929 - 14761049 on Build Release_6_plus_ISO1_MT
          Chromosome Location:
          Chr.3L: 14747929 - 14757962 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0013263
          FBgn0260233
          Assay ID Dm01798607_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001038924.4 NP_001034013.2 1 - 2 427 91
          NM_001038925.3 NP_001034014.1 2 - 3 476 91
          NM_001038926.2 NP_001034015.1 2 - 3 542 91
          NM_001274917.1 NP_001261846.1 2 - 3 542 91
          NM_206355.4 NP_996077.1 2 - 3 542 91
          NM_206356.3 NP_996078.1 2 - 3 542 91
          NM_206357.3 NP_996079.1 2 - 3 900 91
          NM_206358.4 NP_996080.1 2 - 3 717 91
          NM_206359.4 NP_996081.1 2 - 3 721 91
          NM_206360.1 NP_996082.1 3 - 4 845 91
          GenBank mRNA AY069651.1 - 2 - 3 700 91
          Gene Symbol: CG42507
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001169972.2 NP_001163443.1 2 - 3 542 91
          NM_001169973.2 NP_001163444.1 2 - 3 542 91

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_001169972.2, NM_001169973.2 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear division
          chromatin organization
          regulation of transcription from RNA polymerase II promoter
          spermatogenesis
          imaginal disc-derived wing morphogenesis
          dosage compensation
          chromatin assembly
          positive regulation of chromatin silencing
          nucleosome organization
          response to ecdysone
          syncytial blastoderm mitotic cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          oogenesis
          positive regulation of phosphorylation of RNA polymerase II C-terminal domain
          biological_process
          bacterial-type RNA polymerase transcriptional activator activity, sequence-specific DNA binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          POZ domain binding
          identical protein binding
          protein homodimerization activity
          sequence-specific DNA binding
          protein heterodimerization activity
          molecular_function

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline