Search
Search

Trf2| Gene Symbol: Trf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103435.3 | NP_001096905.1 | 8 - 9 | 5271 | 54 | |
| NM_001169230.2 | NP_001162701.1 | 7 - 8 | 5190 | 54 | ||
| NM_001272400.1 | NP_001259329.1 | 8 - 9 | 5240 | 54 | ||
| NM_001272401.1 | NP_001259330.1 | 6 - 7 | 4980 | 54 | ||
| NM_078529.4 | NP_511084.2 | 6 - 7 | 5074 | 54 | ||
| NM_206654.2 | NP_996377.1 | 8 - 9 | 5365 | 54 | ||
| GenBank mRNA | AY069554.1 | - | 3 - 4 | 2645 | 54 | |
| DQ162845.1 | - | 8 - 9 | 5251 | 54 | ||
Set Membership: |
|