Search
Search

Prosbeta4R1| Gene Symbol: Prosbeta4R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134854.3 | NP_608698.2 | 1 - 2 | 190 | 57 | |
| GenBank mRNA | AY075275.1 | - | 1 - 2 | 124 | 57 | |
Set Membership: |
|