Search
Search

CG43707| Gene Symbol: CG43707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273075.1 | NP_001260004.1 | 7 - 8 | 2280 | 64 | |
| NM_001273076.1 | NP_001260005.1 | 7 - 8 | 1628 | 64 | ||
| NM_001273078.1 | NP_001260007.1 | 7 - 8 | 1628 | 64 | ||
| NM_134962.3 | NP_608806.3 | 2 - 3 | 477 | 64 | ||
Set Membership: |
|