Search
Search

mRpL28| Gene Symbol: mRpL28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273134.1 | NP_001260063.1 | 2 - 3 | 1006 | 121 | |
| NM_135043.4 | NP_608887.1 | 2 - 3 | 1006 | 121 | ||
| GenBank mRNA | BT030944.1 | - | 2 - 3 | 957 | 121 | |
Set Membership: |
|