Search
Search

Baldspot| Gene Symbol: Baldspot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140652.4 | NP_648909.1 | 3 - 4 | 520 | 66 | |
| NM_168678.3 | NP_730187.2 | 3 - 4 | 580 | 66 | ||
| GenBank mRNA | AF265296.1 | - | 3 - 4 | 532 | 66 | |
| AY061121.1 | - | 3 - 4 | 532 | 66 | ||
Set Membership: |
|