Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140945.3 | NP_649202.1 | 2 - 3 | 307 | 69 | |
GenBank mRNA | BT023262.2 | - | 2 - 3 | 270 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |