Search
Search

dgt2| Gene Symbol: dgt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298927.1 | NP_001285856.1 | 1 - 2 | 321 | 83 | |
| NM_135687.5 | NP_609531.1 | 1 - 2 | 321 | 83 | ||
| GenBank mRNA | AY070785.1 | - | 1 - 2 | 293 | 83 | |
Set Membership: |
|