Search
Search

sws| Gene Symbol: sws | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078520.3 | NP_511075.3 | 10 - 11 | 4595 | 51 | |
| NM_167140.2 | NP_727224.2 | 10 - 11 | 4595 | 51 | ||
| NM_167141.3 | NP_727225.1 | 3 - 4 | 952 | 51 | ||
| GenBank mRNA | BT133162.1 | - | 10 - 11 | 4595 | 51 | |
Set Membership: |
|