Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: hiw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298329.1 | NP_001285258.1 | 16 - 17 | 11405 | 81 | |
NM_078604.4 | NP_511159.3 | 16 - 17 | 11405 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |