Search
Search

kis| Gene Symbol: kis | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144289.2 | NP_001137761.1 | 8 - 9 | 9249 | 64 | |
| NM_001258891.2 | NP_001245820.1 | 8 - 9 | 9430 | 64 | ||
| NM_078717.3 | NP_523441.1 | 8 - 9 | 9249 | 64 | ||
| GenBank mRNA | AF215703.1 | - | 8 - 9 | 9221 | 64 | |
| MN044634.1 | - | 8 - 9 | 9294 | 64 | ||
Set Membership: |
|