Search
Search

eve| Gene Symbol: eve | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078946.4 | NP_523670.2 | 1 - 2 | 232 | 73 | |
| GenBank mRNA | BT128763.1 | - | 1 - 2 | 212 | 73 | |
| X05138.1 | - | 310 | 73 | |||
Set Membership: |
|