Search
Search

Cals| Gene Symbol: Cals | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272157.1 | NP_001259086.1 | 7 - 8 | 2156 | 121 | |
| NM_001297793.1 | NP_001284722.1 | 7 - 8 | 2159 | 121 | ||
| NM_079893.3 | NP_524632.1 | 7 - 8 | 2156 | 121 | ||
| NM_166823.2 | NP_726646.1 | 7 - 8 | 2159 | 121 | ||
| GenBank mRNA | AJ289018.1 | - | 7 - 8 | 2122 | 121 | |
| AY121644.1 | - | 7 - 8 | 2124 | 121 | ||
Set Membership: |
|