Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEAF-32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079023.5 | NP_523747.1 | 1 - 2 | 361 | 72 | |
GenBank mRNA | X86007.1 | - | 439 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |