Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: qkr58E-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299793.1 | NP_001286722.1 | 3 - 4 | 984 | 86 | |
NM_001299794.1 | NP_001286723.1 | 3 - 4 | 984 | 86 | ||
NM_079085.5 | NP_523809.2 | 3 - 4 | 984 | 86 | ||
GenBank mRNA | AY118745.1 | - | 3 - 4 | 1008 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |