Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bol | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169923.3 | NP_001163394.1 | 8 - 9 | 1380 | 76 | |
NM_001274685.2 | NP_001261614.1 | 6 - 7 | 1324 | 76 | ||
NM_001300063.1 | NP_001286992.1 | 6 - 7 | 1728 | 76 | ||
NM_079265.4 | NP_523989.1 | 6 - 7 | 1309 | 76 | ||
NM_168318.3 | NP_729456.1 | 7 - 8 | 1158 | 76 | ||
NM_168319.2 | NP_729457.1 | 7 - 8 | 1158 | 76 | ||
NM_168320.2 | NP_729458.1 | 7 - 8 | 1651 | 76 | ||
GenBank mRNA | BT011495.1 | - | 8 - 9 | 1346 | 76 | |
U51858.1 | - | 6 - 7 | 1299 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |