Search
Search

Hsp23| Gene Symbol: Hsp23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259765.2 | NP_001246694.1 | 1 | 754 | 144 | |
| NM_079275.4 | NP_523999.1 | 1 | 754 | 144 | ||
| GenBank mRNA | AY061081.1 | - | 1 | 725 | 144 | |
Set Membership: |
|