Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Uch-L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274703.1 | NP_001261632.1 | 1 - 2 | 192 | 109 | |
NM_079279.3 | NP_524003.1 | 1 - 2 | 192 | 109 | ||
GenBank mRNA | AF132567.1 | - | 1 - 2 | 174 | 109 | |
AF145312.1 | - | 1 - 2 | 74 | 109 | ||
BT001294.1 | - | 718 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |