Search
Search

LanB2| Gene Symbol: LanB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300080.1 | NP_001287009.1 | 4 - 5 | 807 | 63 | |
| NM_079282.2 | NP_524006.1 | 4 - 5 | 712 | 63 | ||
| GenBank mRNA | BT021394.1 | - | 4 - 5 | 687 | 63 | |
Set Membership: |
|