Search
Search

apolpp| Gene Symbol: apolpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272156.1 | NP_001259085.1 | 3 - 4 | 250 | 66 | |
| NM_001297791.1 | NP_001284720.1 | 3 - 4 | 301 | 66 | ||
| NM_001297792.1 | NP_001284721.1 | 3 - 4 | 375 | 66 | ||
| NM_079895.3 | NP_524634.2 | 3 - 4 | 250 | 66 | ||
Set Membership: |
|