Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rbfox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104102.4 | NP_001097572.1 | 9 - 10 | 2652 | 77 | |
NM_001104103.3 | NP_001097573.1 | 9 - 10 | 2637 | 77 | ||
NM_001259778.1 | NP_001246707.1 | 11 - 12 | 3400 | 77 | ||
NM_001259779.1 | NP_001246708.1 | 9 - 10 | 3160 | 77 | ||
NM_168412.4 | NP_729614.2 | 10 - 11 | 2835 | 77 | ||
NM_168413.3 | NP_729615.2 | 11 - 12 | 2877 | 77 | ||
GenBank mRNA | BT010058.1 | - | 9 - 10 | 2624 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |