Search
Search

brm| Gene Symbol: brm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274977.1 | NP_001261906.1 | 2 - 3 | 371 | 78 | |
| NM_080498.3 | NP_536746.1 | 2 - 3 | 371 | 78 | ||
| NM_168641.1 | NP_730089.1 | 1 - 2 | 585 | 78 | ||
| GenBank mRNA | AY095048.1 | - | 2 - 3 | 369 | 78 | |
Set Membership: |
|