Search
Search

Cpr11B| Gene Symbol: Cpr11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272556.1 | NP_001259485.1 | 1 - 2 | 386 | 62 | |
| NM_132579.3 | NP_572807.1 | 1 - 2 | 386 | 62 | ||
| GenBank mRNA | AY060775.1 | - | 1 - 2 | 369 | 62 | |
Set Membership: |
|