Search
Search

Blimp-1| Gene Symbol: Blimp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274513.1 | NP_001261442.1 | 2 - 3 | 1040 | 65 | |
| NM_001274514.1 | NP_001261443.1 | 2 - 3 | 1045 | 65 | ||
| NM_001274515.1 | NP_001261444.1 | 2 - 3 | 873 | 65 | ||
| NM_001274516.1 | NP_001261445.1 | 2 - 3 | 610 | 65 | ||
| NM_001300016.1 | NP_001286945.1 | 2 - 3 | 1040 | 65 | ||
| NM_139725.3 | NP_647982.1 | 2 - 3 | 1040 | 65 | ||
| GenBank mRNA | AY071225.1 | - | 2 - 3 | 1040 | 65 | |
Set Membership: |
|