Search
Search

Mp20| Gene Symbol: Mp20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014522.2 | NP_001014522.2 | 2 - 3 | 441 | 104 | |
| NM_057295.4 | NP_476643.1 | 2 - 3 | 441 | 104 | ||
| GenBank mRNA | AY071496.1 | - | 2 - 3 | 438 | 104 | |
Set Membership: |
|