Search
Search

sgl| Gene Symbol: sgl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057632.4 | NP_476980.1 | 1 - 2 | 364 | 61 | |
| GenBank mRNA | AF000570.1 | - | 1 - 2 | 366 | 61 | |
| AF007870.1 | - | 1 - 2 | 374 | 61 | ||
| AF009013.1 | - | 1 - 2 | 360 | 61 | ||
| AY052137.1 | - | 1 - 2 | 347 | 61 | ||
Set Membership: |
|