Search
Search

RpL11| Gene Symbol: RpL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299685.1 | NP_001286614.1 | 4 - 5 | 504 | 72 | |
| NM_057706.5 | NP_477054.1 | 4 - 5 | 504 | 72 | ||
| GenBank mRNA | AY094790.1 | - | 3 - 4 | 468 | 72 | |
Set Membership: |
|