Search
Search

Thor| Gene Symbol: Thor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298659.1 | NP_001285588.1 | 1 - 2 | 237 | 60 | |
| NM_057947.4 | NP_477295.1 | 1 - 2 | 325 | 60 | ||
| GenBank mRNA | AF132557.1 | - | 1 - 2 | 304 | 60 | |
Set Membership: |
|