Search
Search

gbb| Gene Symbol: gbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299857.1 | NP_001286786.1 | 2 | 1473 | 137 | |
| NM_057992.3 | NP_477340.1 | 1 | 1554 | 137 | ||
| GenBank mRNA | M84795.1 | - | 1 | 1502 | 135 | |
Set Membership: |
|