Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Src64B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014561.2 | NP_001014561.1 | 4 - 5 | 1508 | 83 | |
NM_001014562.3 | NP_001014562.1 | 3 - 4 | 1260 | 83 | ||
NM_001014563.3 | NP_001014563.1 | 5 - 6 | 1649 | 83 | ||
NM_001014564.3 | NP_001014564.1 | 5 - 6 | 1410 | 83 | ||
NM_001202122.1 | NP_001189051.1 | 4 - 5 | 1203 | 83 | ||
NM_001259699.2 | NP_001246628.1 | 5 - 6 | 1410 | 83 | ||
NM_001259700.2 | NP_001246629.1 | 5 - 6 | 1410 | 83 | ||
NM_001300008.1 | NP_001286937.1 | 5 - 6 | 1475 | 83 | ||
NM_080195.4 | NP_524934.2 | 5 - 6 | 1475 | 83 | ||
GenBank mRNA | AY051781.1 | - | 5 - 6 | 1452 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |